1-Methylpiperidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 626-67-5
Molecular Formula: C6H13N
Formula Weight: 99.18
Flash Point: 3°(37°F)
Refractive Index: 1.4378
Odor: Amine-like
pH: 12
Melting Point: -50°
Boiling Point: 105-107°
Beilstein Reference: 1073
UN No.: 2399
MDL No.: MFCD00006491
Formula:
C₆H₁₃N Boiling Pt: 105…107 °C Melting Pt: –50 °C Density: 0.816 Flash Pt: 3 °C (37 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006491 CAS Number: 626-67-5 EINECS: 210-959-9 UN: 2399 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...