(±)-2-Methyl-2,4-pentanediol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexylene glycol, (±)-2,4-Dihydroxy-2-methyl pentane, (±)-2,4-Dihydroxy-2-methyl pentane, 2-Methylpentane-2,4-diol, Hexylene Glycol

L03187-0F L03187-AE L03187-AP
AAAL03187-0FEA 70.9 USD
AAAL03187-0F AAAL03187-AE AAAL03187-AP
(±)-2-Methyl-2,4-pentanediol 98%
(±)-2-Methyl-2,4-pentanediol

Purity: 98% CAS No.: 107-41-5 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 93°(199°F) Refractive Index: 1.4270 Odor: Weak Melting Point: -40° Boiling Point: 197° Beilstein Reference: 1098298 Merck Reference: 14,4710 MDL No.: MFCD00004547

Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 198
Melting Pt: -40
Density: 0.922
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004547
CAS Number: 107-41-5
EINECS: 203-489-0
Merck Index: 14,4710

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR