Perylene 98+%

Supplier: Thermo Scientific Chemicals

L03047-03 L03047-06
AAAL03047-03EA 48 USD
AAAL03047-03 AAAL03047-06
Perylene 98+%
Perylene

Purity: 98+% CAS No.: 198-55-0 Molecular Formula: C20H12 Formula Weight: 252.32 Odor: Odorless Melting Point: 276-280° Beilstein Reference: 1911335 Merck Reference: 14,7184 MDL No.: MFCD00004142

Formula: C₂₀H₁₂
MW: 252.32 g/mol
Boiling Pt: 443
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00004142
CAS Number: 198-55-0
EINECS: 205-900-9
Merck Index: 14,7184

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR