Thiocarbohydrazide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
TCH
L01205-14
L01205-22
L01205-06
AAAL01205-14EA
141
USD
AAAL01205-14
AAAL01205-22
AAAL01205-06
Thiocarbohydrazide 97%
Thiocarbohydrazide
Purity: 97%
CAS No.: 2231-57-4
Molecular Formula: CH6N4S
Formula Weight: 106.15
Storage Temperature: 2°C to 8°C
Melting Point: ca 175° dec.
Beilstein Reference: 506657
UN No.: 2811
MDL No.: MFCD00007616
Formula:
CH₆N₄S Melting Pt: ca 175 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007616 CAS Number: 2231-57-4 EINECS: 218-769-8 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...