Glutarimide 98%
Supplier: Thermo Scientific Chemicals
L00968-14
L00968-06
L00968-22
AAAL00968-14EA
172
USD
AAAL00968-14
AAAL00968-06
AAAL00968-22
Glutarimide 98%
Glutarimide
Purity: 98%
CAS No.: 1121-89-7
Molecular Formula: C5H7NO2
Formula Weight: 113.12
Odor: Nearly odorless
Melting Point: 153-157°
Beilstein Reference: 110052
MDL No.: MFCD00006670
Formula:
C₅H₇NO₂ Melting Pt: 153…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006670 CAS Number: 1121-89-7 EINECS: 214-340-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...