Triphenylene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9,10-Benzophenanthrene, Isochrysene

L00955-06 L00955-03
AAAL00955-06EA 264 USD
AAAL00955-06 AAAL00955-03
Triphenylene 98%
Triphenylene

Purity: 98% CAS No.: 217-59-4 Molecular Formula: C18H12 Formula Weight: 228.30 Melting Point: 194-199° Boiling Point: 438° Merck Reference: 14,9740 MDL No.: MFCD00001108

Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 438
Melting Pt: 199
Density: 1.302
Storage Temperature: Ambient
MDL Number: MFCD00001108
CAS Number: 217-59-4
EINECS: 205-922-9
UN: 3077
ADR: 9 III
Merck Index: 14,9740

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR