4-(3-Chlorophenoxy)piperidine 98%

Supplier: Thermo Scientific Chemicals
Warning

H50956-03
AAAH50956-03EA 292 USD
AAAH50956-03 AAAH50956-MD
4-(3-Chlorophenoxy)piperidine 98%
4-(3-Chlorophenoxy)piperidine

Purity: 98% CAS No.: 97840-40-9 Molecular Formula: C11H14ClNO Formula Weight: 211.69 MDL No.: MFCD08061087

Formula: C₁₁H₁₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD08061087
CAS Number: 97840-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR