4-Chloro-6-(trifluoromethyl)quinoline 99%

Supplier: Thermo Scientific Chemicals
Warning

H50539-06 H50539-03
AAAH50539-06EA 270 USD
AAAH50539-06 AAAH50539-03
4-Chloro-6-(trifluoromethyl)quinoline 99%
4-Chloro-6-(trifluoromethyl)quinoline

Purity: 99% CAS No.: 49713-56-6 Molecular Formula: C10H5ClF3N Formula Weight: 231.61 MDL No.: MFCD00153106

Formula: C₁₀H₅ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD00153106
CAS Number: 49713-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR