3,5-Dichloro-4-hydroxybenzonitrile 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloroxynil

H50432-03
AAAH50432-03EA 130 USD
AAAH50432-03 AAAH50432-06
3,5-Dichloro-4-hydroxybenzonitrile 97+%
3,5-Dichloro-4-hydroxybenzonitrile

Purity: 97+% CAS No.: 1891-95-8 Molecular Formula: C7H3Cl2NO Formula Weight: 188.01 Melting Point: 143-144° UN No.: 3439 MDL No.: MFCD00002177

Formula: C₇H₃Cl₂NO
Melting Pt: 143…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00002177
CAS Number: 1891-95-8
EINECS: 217-572-4
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR