1-Bromo-3,5-dichlorobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dichlorobromobenzene

B25492-06 B25492-14 B25492-22
AAAB25492-06EA 23.6 USD
AAAB25492-06 AAAB25492-14 AAAB25492-22
1-Bromo-3,5-dichlorobenzene 97%
1-Bromo-3,5-dichlorobenzene

Purity: 97% CAS No.: 19752-55-7 Molecular Formula: C6H3BrCl2 Formula Weight: 225.91 Melting Point: 73-77° Boiling Point: 232° MDL No.: MFCD00000584

Formula: C₆H₃BrCl₂
Boiling Pt: 232 °C
Melting Pt: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR