2,7-Dichlorofluorene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,7-Dichloro-9H-fluorene

B24717-06
AAAB24717-06EA 120 USD
AAAB24717-06 AAAB24717-03
2,7-Dichlorofluorene 97%
2,7-Dichlorofluorene

Purity: 97% CAS No.: 7012-16-0 Molecular Formula: C13H10Cl2 Formula Weight: 235.12 Melting Point: 300° dec. MDL No.: MFCD00032840

Formula: C₁₃H₈Cl₂
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00032840
CAS Number: 7012-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR