4-Ethylbenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Ethylbenzene-1-sulfonyl chloride, 4-Ethylbenzene-1-sulphonyl chloride

B24682-06 B24682-14
AAAB24682-22EA 621.43 USD
AAAB24682-22 AAAB24682-06 AAAB24682-14
4-Ethylbenzenesulfonyl chloride 97%
4-Ethylbenzenesulphonyl chloride

Purity: 97% CAS No.: 16712-69-9 Molecular Formula: C8H9ClO2S Formula Weight: 204.68 Refractive Index: 1.5450 Melting Point: 8-12° Boiling Point: 134-136°/7mm Beilstein Reference: 2090130 UN No.: 3265 MDL No.: MFCD00041480

Formula: C₈H₉ClO₂S
Boiling Pt: 134…136 °C/7 mm
Melting Pt: 8…12 °C
Density: 1.268
Storage Temperature: Ambient
MDL Number: MFCD00041480
CAS Number: 16712-69-9
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR