Dibutylbis(2,4-pentanedionato-O,O')tin 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Di-n-butyltinbis(acetylacetonate), Di-n-butyltin bis(2,4-pentanedionate), (OC-6-11)-Dibutylbis(2,4-pentanedionato-κO2,κO4)Tin

B23921-0B B23921-30
AAAB23921-0BEA 286 USD
AAAB23921-0B AAAB23921-30
Dibutylbis(2,4-pentanedionato-O,O')tin 95%
Dibutylbis(2,4-pentanedionato-O,O')tin

Purity: 95% CAS No.: 22673-19-4 Molecular Formula: C18H32O4Sn Formula Weight: 431.16 Flash Point: 40°(104°F) Refractive Index: 1.5150 Odor: Weak Melting Point: 26° Boiling Point: 150° UN No.: 1992 MDL No.: MFCD00077994

Formula: C₁₈H₃₂O₄Sn
Boiling Pt: 150 °C
Melting Pt: 26 °C
Density: 1.22
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077994
CAS Number: 22673-19-4
EINECS: 245-152-0
UN: 1992
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR