Catechol violet

Supplier: Thermo Scientific Chemicals

B23513-06 B23513-14
AAAB23513-06EA 64.53 USD
AAAB23513-06 AAAB23513-14
Catechol violet
Catechol violet

CAS No.: 115-41-3 Molecular Formula: C19H14O7S Formula Weight: 386.38 Beilstein Reference: 353938 MDL No.: MFCD00005868

Formula: C₁₉H₁₄O₇S
Storage Temperature: Ambient
MDL Number: MFCD00005868
CAS Number: 115-41-3
EINECS: 204-088-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR