Benzyl methyl ether 97%

Supplier: Thermo Scientific Chemicals
Warning

B23490-09 B23490-18
AAAB23490-09EA 41.3 USD
AAAB23490-09 AAAB23490-18 AAAB23490-30
Benzyl methyl ether 97%
Benzyl methyl ether

Purity: 99% CAS No.: 538-86-3 Molecular Formula: C6H5CH2OCH3 Formula Weight: 122.17 Flash Point: 56°(133°F) Refractive Index: 1.5015 Melting Point: -53° Boiling Point: 173-175° Merck Reference: 14,1140 UN No.: 3271 MDL No.: MFCD00025901

Formula: C₈H₁₀O
Boiling Pt: 173…175 °C
Melting Pt: –53 °C
Density: 0.940
Flash Pt: 56 °C(133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00025901
CAS Number: 538-86-3
EINECS: 208-705-7
UN: 3271
ADR: "3",III
Merck Index: 14,1140

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR