Phenylphosphonic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenylphosphonic Acid
B22939-22
AAAB22939-22EA
69.9
USD
AAAB22939-22
AAAB22939-36
Phenylphosphonic acid 98%
Phenylphosphonic acid
Purity: 98%
CAS No.: 1571-33-1
Molecular Formula: C6H7O3P
Formula Weight: 158.09
Melting Point: 160-162°
Beilstein Reference: 2245168
UN No.: 3261
MDL No.: MFCD00002136
Formula:
C₆H₇O₃P Melting Pt: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002136 CAS Number: 1571-33-1 EINECS: 216-388-1 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...