Diisopropyl cyanomethylphosphonate 97%

Supplier: Thermo Scientific Chemicals
Danger

B22163-06 B22163-14
AAB22163-06EA 80.7 USD
AAB22163-06 AAAB22163-14
Diisopropyl cyanomethylphosphonate 97%
Diisopropyl cyanomethylphosphonate

Purity: 97% CAS No.: 21658-95-7 Molecular Formula: C8H16NO3P Formula Weight: 205.19 Flash Point: 110°(230°F) Refractive Index: 1.4295 Boiling Point: 108-110°/0.08m Beilstein Reference: 1946176 UN No.: 2927 MDL No.: MFCD00009812

Formula: C₈H₁₆NO₃P
Boiling Pt: 108…110 °C/0.08 mm
Density: 1.039
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009812
CAS Number: 21658-95-7
EINECS: 244-511-9
UN: 2927
ADR: "6.1",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR