Guanidinium thiocyanate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
B21250-22
B21250-36
B21250-0E
AAB21250-22EA
80.45
USD
AAB21250-22
AAB21250-36
AAAB21250-0E
Guanidinium thiocyanate 99%
Guanidinium thiocyanate
Purity: 99%
CAS No.: 593-84-0
Molecular Formula: CH5N3.HSCN
Formula Weight: 118.16
Melting Point: 118-122°
Beilstein Reference: 3563461
Formula:
CH₅N₃·CHNS Melting Pt: 118…122 °C Density: 1.29 Storage Temperature: Ambient |
MDL Number:
MFCD00013027 CAS Number: 593-84-0 EINECS: 209-812-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...