Triethyl borate 97%

Supplier: Thermo Scientific Chemicals
Danger

A18728-AE A18728-AP A18728-0F
AAAA18728-AEEA 48.3 USD
AAAA18728-AE AAAA18728-AP AAAA18728-0F
Triethyl borate 97%
Triethyl borate

Purity: 97% CAS No.: 150-46-9 Molecular Formula: C6H15BO3 Formula Weight: 145.99 Flash Point: 11°(51°F) Refractive Index: 1.3740 Odor: Mild Melting Point: -85° Boiling Point: 117-118° Beilstein Reference: 1699052 UN No.: 1176 MDL No.: MFCD00009073

Formula: C₆H₁₅BO₃
Boiling Pt: 117…118 °C
Melting Pt: –85 °C
Density: 0.858
Flash Pt: 11 °C (51 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012846
CAS Number: 150-46-9
EINECS: 205-760-9
UN: 1176
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR