(3-Glycidoxypropyl)trimethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Glycidyloxypropyl)trimethoxysilane, 3-Glycidoxypropyltrimethoxysilane, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilane

A18431-22 A18431-36 A18431-14
AAAA18431-22EA 72.55 USD
AAAA18431-22 AAAA18431-36 AAAA18431-14
(3-Glycidoxypropyl)trimethoxysilane 97%
(3-Glycidoxypropyl)trimethoxysilane

Purity: 97% CAS No.: 2530-83-8 Molecular Formula: C9H20O5Si Formula Weight: 236.34 Flash Point: 135°(275°F) Refractive Index: 1.4290 Odor: Odorless Melting Point: -70° Boiling Point: 119-121°/2mm Beilstein Reference: 4308125 MDL No.: MFCD00005144

Formula: C₉H₂₀O₅Si
MW: 236.34 g/mol
Boiling Pt: 299
Melting Pt: 0
Density: 1.072
Flash Pt: 135 °C (275 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005144
CAS Number: 2530-83-8
EINECS: 219-784-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR