Sodium hydroxide 98%, soluble
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
A18395-0E
A18395-36
A18395-0C
AAAA18395-0EEA
171.28
USD
AAAA18395-0E
AAAA18395-36
AAAA18395-0C
Sodium hydroxide 98%, soluble
Sodium hydroxide
Purity: 98%
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Odor: Odorless
pH: 14
Melting Point: 318°
Boiling Point: 1390°
Merck Reference: 14,8627
UN No.: 1823
MDL No.: MFCD00003548
Formula:
NaOH Boiling Pt: 1390 °C Melting Pt: 318 °C Density: 2.13 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,8627 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...