1-Butanesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Butanesulphonylchloride, 1-Butanesulfonylchloride
A18295-22
A18295-06
AAAA18295-22EA
256
USD
AAAA18295-22
AAA18295-06
1-Butanesulfonyl chloride 98%
1-Butanesulphonyl chloride
Purity: 98%
CAS No.: 2386-60-9
Molecular Formula: C4H9ClO2S
Formula Weight: 156.63
Flash Point: 79°(174°F)
Refractive Index: 1.4560
Melting Point: -29°
Boiling Point: 80-81°/9mm
Beilstein Reference: 1748742
UN No.: 3265
MDL No.: MFCD00007464
Formula:
C₄H₉ClO₂S MW: 156.63 g/mol Boiling Pt: 208 Melting Pt: -25 Density: 1.210 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007464 CAS Number: 2386-60-9 EINECS: 219-204-8 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...