Sodium carbonate decahydrate 99+%

Supplier: Thermo Scientific Chemicals
Warning

A17484-0C A17484-36 A17484-0E
AAAA17484-0CEA 453 USD
AAAA17484-0C AAAA17484-36 AAAA17484-0E
Sodium carbonate decahydrate 99+%
Sodium carbonate decahydrate

Purity: 99+% CAS No.: 6132-02-1 Molecular Formula: CNa2O3.10H2O Formula Weight: 286.14 (105.99anhy) Melting Point: 32-34° Merck Reference: 14,8596 MDL No.: MFCD00149178

Formula: Na₂CO₃·10H₂O
Melting Pt: 32…34 °C
Density: 1.46
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1
EINECS: 207-838-8
Merck Index: 14,8596

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR