Ethylene glycol monomethyl ether (2-methoxyethanol) 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethylene glycol monomethyl ether
A17459-0D
A17459-AP
A17459-0F
AAAA17459-0DEA
275
USD
AAAA17459-0D
AAAA17459-AP
AAAA17459-0F
Ethylene glycol monomethyl ether (2-methoxyethanol) 99%
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Solvent for ninhydrin color reagent
Purity: 99%
CAS No.: 109-86-4
Molecular Formula: C3H8O2
Formula Weight: 76.10
Flash Point: 46°(115°F)
Refractive Index: 1.4025
Odor: Characteristic
pH: 4-7
Melting Point: -85°
Boiling Point: 124-125°
Beilstein Reference: 1731074
Merck Reference: 14,6038
UN No.: 1188
MDL No.: MFCD00002867
Formula:
C₃H₈O₂ Boiling Pt: 124…125 °C Melting Pt: –85 °C Density: 0.965 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002867 CAS Number: 109-86-4 EINECS: 203-713-7 UN: 1188 ADR: 3,III Merck Index: 14,6038 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...