Ethylene glycol monomethyl ether (2-methoxyethanol) 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethylene glycol monomethyl ether

A17459-0D A17459-AP A17459-0F
AAAA17459-0DEA 275 USD
AAAA17459-0D AAAA17459-AP AAAA17459-0F
Ethylene glycol monomethyl ether (2-methoxyethanol) 99%
Ethylene glycol monomethyl ether (2-Methoxyethanol)

Solvent for ninhydrin color reagent


Purity: 99% CAS No.: 109-86-4 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: 46°(115°F) Refractive Index: 1.4025 Odor: Characteristic pH: 4-7 Melting Point: -85° Boiling Point: 124-125° Beilstein Reference: 1731074 Merck Reference: 14,6038 UN No.: 1188 MDL No.: MFCD00002867

Formula: C₃H₈O₂
Boiling Pt: 124…125 °C
Melting Pt: –85 °C
Density: 0.965
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 14,6038

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR