Methyl red sodium salt ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate

A17455-36 A17455-14 A17455-22
AAAA17455-36EA 261 USD
AAAA17455-36 AAAA17455-14 AAAA17455-22
Methyl red sodium salt ≥98%
Methyl red sodium salt

CAS No.: 845-10-3 Molecular Formula: C15H14N3NaO2 Formula Weight: 291.29 Beilstein Reference: 4065488 MDL No.: MFCD00002426

Formula: C₁₅H₁₄N₃NaO₂
MW: 291.28 g/mol
Melting Pt: > 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002426
CAS Number: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR