Diammonium hydrogen orthophosphate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

A17416-36
AAAA17416-36EA 70 USD
AAAA17416-36
Diammonium hydrogen orthophosphate 98%
di-Ammonium hydrogen orthophosphate

Purity: 98% CAS No.: 7783-28-0 Molecular Formula: H9N2O4P Formula Weight: 132.06 Melting Point: 155° dec. Merck Reference: 14,542 MDL No.: MFCD00010891

Formula: (NH₄)₂HPO₄
Melting Pt: 155 °C (dec.)
Density: 1.619
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 14,542

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR