Strontium chloride hexahydrate 99+%

Supplier: Thermo Scientific Chemicals
Danger

A16764-30 A16764-0B
AAAA16764-30EA 87 USD
AAAA16764-30 AAAA16764-0B
Strontium chloride hexahydrate 99+%
Strontium chloride hexahydrate

Purity: 99+% CAS No.: 10025-70-4 Molecular Formula: SrCl2.6H2O Formula Weight: 266.62 (158.53anhy) Refractive Index: 1.650 Merck Reference: 14,8840 MDL No.: MFCD00149865

Formula: SrCl₂·6H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 61 °C
Density: 1.93
Storage Temperature: Ambient
MDL Number: MFCD00149865
CAS Number: 10025-70-4
EINECS: 233-971-6
Merck Index: 14,8840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR