Copper(II) bromide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cupric bromide, Cuprous bromide
A16335-14
A16335-22
A16335-36
AAAA16335-14EA
36.43
USD
AAAA16335-14
AAAA16335-22
AAAA16335-36
Copper(II) bromide 99%
Copper (II) bromide
Purity: 99%
CAS No.: 7789-45-9
Molecular Formula: CuBr2
Formula Weight: 223.36
Odor: Odorless
Melting Point: 498°
Boiling Point: 900°
Merck Reference: 14,2629
UN No.: 3260
MDL No.: MFCD00010970
Formula:
Br₂Cu MW: 223.35 g/mol Boiling Pt: 900 Melting Pt: 498 Density: 4.710 Storage Temperature: Ambient |
MDL Number:
MFCD00010970 CAS Number: 7789-45-9 EINECS: 232-167-2 UN: 3260 ADR: 8 III Merck Index: 14,2629 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...