Potassium iodate 98%

Supplier: Thermo Scientific Chemicals
Danger

A16162-36 A16162-22 A16162-0E
AAA16162-36EA 203 USD
AAA16162-36 AAA16162-22 AAAA16162-0E
Potassium iodate 98%
Potassium iodate

Purity: 98% CAS No.: 7758-05-6 Molecular Formula: KIO3 Formula Weight: 214.00 Melting Point: 560° dec. Merck Reference: 14,7642 MDL No.: MFCD00011406

Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560 (dec.)
Density: 3.89
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1 III
Merck Index: 14,7642

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR