Benzenesulfonic acid 94%

Supplier: Thermo Scientific Chemicals
Danger

A16148-36 A16148-22 A16148-0E
AAAA16148-36EA 39 USD
AAAA16148-36 AAA16148-22 AAAA16148-0E
Benzenesulfonic acid 94%
Benzenesulphonic acid

Acid catalyst for direct esterification of amino acids and peptides


Purity: 94% CAS No.: 98-11-3 Molecular Formula: C6H6O3S Formula Weight: 158.18 Flash Point: 110°(230°F) Melting Point: ca 45-50° Beilstein Reference: 742513 Merck Reference: 14,1070 UN No.: 2585 MDL No.: MFCD00011689

Formula: C₆H₆O₃S
MW: 158.18 g/mol
Boiling Pt: 319
Melting Pt: 50
Density: 1.32
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011689
CAS Number: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: 8 III
Merck Index: 14,1070

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR