Copper(II) chloride dihydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

A16064-0B A16064-30
AAAA16064-0BEA 197.58 USD
AAAA16064-0B AAAA16064-30
Copper(II) chloride dihydrate 99%
Copper (II) chloride dihydrate

Purity: 99% CAS No.: 10125-13-0 Molecular Formula: CuCl2.2H2O Formula Weight: 170.48 (134.45anhy) Odor: Odorless pH: 3.0-3.8 Melting Point: 100° dec. Merck Reference: 14,2633 UN No.: 2802 MDL No.: MFCD00149674

Formula: CuCl₂·2H₂O
Melting Pt: 100 °C (dec.)
Density: 2.510
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 14,2633

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR