Potassium hexacyanoferrate(II) trihydrate 98+%
Supplier: Thermo Scientific Chemicals
|
|
A15736-0E
A15736-22
A15736-36
AAAA15736-0EEA
296
USD
AAAA15736-0E
AAAA15736-22
AAAA15736-36
Potassium hexacyanoferrate(II) trihydrate 98+%
Potassium hexacyanoferrate (II) trihydrate
Purity: 98+%
CAS No.: 14459-95-1
Molecular Formula: K4Fe(CN)6.3H2O
Formula Weight: 422.39 (368.36anhy)
Odor: Odorless
pH: 9.5
Melting Point: 69-71°
Merck Reference: 14,7631
MDL No.: MFCD00167023
Formula:
K₄Fe(CN)₆·3H₂O Melting Pt: 69…71 °C Density: 1.85 Storage Temperature: Ambient |
MDL Number:
MFCD00167023 CAS Number: 14459-95-1 EINECS: 237-722-2 Merck Index: 14,7631 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...