Pyridinium tribromide tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide
A15684-36
A15684-22
A15684-14
AAAA15684-36EA
349
USD
AAAA15684-36
AAAA15684-22
AAAA15684-14
Pyridinium tribromide tech. 90%, Technical Grade
Pyridinium tribromide
Fieser: 1,967 5,568 6,499 9,399 10,333 15,276
Purity: 90%
CAS No.: 39416-48-3
Molecular Formula: C5H5N.HBr3
Formula Weight: 319.82
Melting Point: 128-136°
Beilstein Reference: 3690144
Merck Reference: 14,7973
UN No.: 3261
MDL No.: MFCD00013223
Formula:
C₅H₆Br₃N MW: 319.82 g/mol Melting Pt: 128…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013223 CAS Number: 39416-48-3 EINECS: 254-446-8 UN: 3261 ADR: 8 III Merck Index: 14,7973 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...