Lanthanum(III) chloride heptahydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

A15575-36 A15575-0E A15575-22
AAAA15575-36EA 312.35 USD
AAAA15575-36 AAAA15575-0E AAAA15575-22
Lanthanum(III) chloride heptahydrate 99%
Lanthanum (III) chloride heptahydrate

Purity: 99% CAS No.: 10025-84-0 Molecular Formula: LaCl3.7H2O Formula Weight: 371.37 (245.27anhy) Odor: Odorless Melting Point: 91° dec. MDL No.: MFCD00149756

Formula: LaCl₃·7H₂O
Boiling Pt: ? 860 °C (1013 hPa)
Melting Pt: 91 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5
Merck Index: 14,5363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR