Furoin 98%
Supplier: Thermo Scientific Chemicals
A15272-06
A15272-14
A15272-22
AAAA15272-06EA
93
USD
AAAA15272-06
AAAA15272-14
AAAA15272-22
Furoin 98%
Furoin
Purity: 98%
CAS No.: 552-86-3
Molecular Formula: C10H8O4
Formula Weight: 192.17
Melting Point: 137-139°
Beilstein Reference: 383995
MDL No.: MFCD00003246
Formula:
C₁₀H₈O₄ Melting Pt: 137…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003246 CAS Number: 552-86-3 EINECS: 209-024-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...