Cetrimonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
A15235-22
A15235-36
A15235-0I
AAA15235-22EA
89.66
USD
AAA15235-22
AAA15235-36
AAAA15235-0I
Cetrimonium bromide 98%
Cetrimonium bromide
Purity: 98%
CAS No.: 57-09-0
Molecular Formula: CH3(CH2)15N(CH3)3Br
Formula Weight: 364.46
Flash Point: 244°(471°F)
Odor: Odorless
pH: 5-7
Melting Point: ca 245° dec.
Beilstein Reference: 3598189
Merck Reference: 14,2025
UN No.: 3077
MDL No.: MFCD00011772
Formula:
CH₃(CH₂)₁₅N(Br)(CH₃)₃ Melting Pt: ∼ 245 °C (dec.) Flash Pt: 244 °C (471 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011772 CAS Number: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9,III Merck Index: 14,2025 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...