1-Chloronaphthalene tech. 85% remainder 2-chloronaphthalene, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 85%
CAS No.: 90-13-1
Molecular Formula: C10H7Cl
Formula Weight: 162.62
Flash Point: 121°(249°F)
Refractive Index: 1.6330
Melting Point: ca -8°
Boiling Point: 259°
Beilstein Reference: 970836
Merck Reference: 14,2149
MDL No.: MFCD00003874
Formula:
C₁₀H₇Cl Boiling Pt: 259 °C Melting Pt: ∼ 8 °C Density: 1.194 Flash Pt: 121 °C (249 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003874 CAS Number: 90-13-1 EINECS: 201-967-3 UN: 3082 ADR: 9,III Merck Index: 14,2149 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...