Cadmium sulfide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14544-0E
A14544-22
A14544-36
AAAA14544-0EEA
580
USD
AAAA14544-0E
AAAA14544-22
AAAA14544-36
Cadmium sulfide 98%
Cadmium sulphide
Purity: 98%
CAS No.: 1306-23-6
Molecular Formula: CdS
Formula Weight: 144.46
Odor: Odorless
Melting Point: 980° subl.
UN No.: 3077
MDL No.: MFCD00010922
Formula:
CdS Melting Pt: 980 °C (subl.) Density: 4.82 Storage Temperature: Ambient |
MDL Number:
MFCD00010922 CAS Number: 1306-23-6 EINECS: 215-147-8 UN: 2570 ADR: 6.1,III Merck Index: 14,1628 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...