Cadmium sulfide 98%

Supplier: Thermo Scientific Chemicals
Danger

A14544-0E A14544-22 A14544-36
AAAA14544-0EEA 580 USD
AAAA14544-0E AAAA14544-22 AAAA14544-36
Cadmium sulfide 98%
Cadmium sulphide

Purity: 98% CAS No.: 1306-23-6 Molecular Formula: CdS Formula Weight: 144.46 Odor: Odorless Melting Point: 980° subl. UN No.: 3077 MDL No.: MFCD00010922

Formula: CdS
Melting Pt: 980 °C (subl.)
Density: 4.82
Storage Temperature: Ambient
MDL Number: MFCD00010922
CAS Number: 1306-23-6
EINECS: 215-147-8
UN: 2570
ADR: 6.1,III
Merck Index: 14,1628

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR