Potassium nitrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14527-36
A14527-0E
A14527-0C
AAAA14527-36EA
56.25
USD
AAAA14527-36
AAAA14527-0E
AAAA14527-0C
Potassium nitrate 99%
Potassium nitrate
Purity: 99%
CAS No.: 7757-79-1
Molecular Formula: KNO3
Formula Weight: 101.11
Odor: Odorless
Melting Point: 334°
Boiling Point: 400° dec.
Merck Reference: 14,7648
UN No.: 1486
MDL No.: MFCD00011409
Formula:
KNO₃ Boiling Pt: 400 °C (dec.) Melting Pt: 334 °C Density: 2.109 Storage Temperature: Ambient |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 14,7648 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...