Potassium nitrate 99%

Supplier: Thermo Scientific Chemicals
Warning

A14527-36 A14527-0E A14527-0C
AAAA14527-36EA 56.25 USD
AAAA14527-36 AAAA14527-0E AAAA14527-0C
Potassium nitrate 99%
Potassium nitrate

Purity: 99% CAS No.: 7757-79-1 Molecular Formula: KNO3 Formula Weight: 101.11 Odor: Odorless Melting Point: 334° Boiling Point: 400° dec. Merck Reference: 14,7648 UN No.: 1486 MDL No.: MFCD00011409

Formula: KNO₃
Boiling Pt: 400 °C (dec.)
Melting Pt: 334 °C
Density: 2.109
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 14,7648

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR