Silicon carbide, coarse powder 46 grit

Supplier: Thermo Scientific Chemicals
Warning

A14470-0C A14470-0E A14470-36
AAAA14470-0CEA 456 USD
AAAA14470-0C AAAA14470-0E AAAA14470-36
Silicon carbide, coarse powder 46 grit
Silicon carbide

CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Odor: Odorless MDL No.: MFCD00049531

Formula: SiC
Melting Pt: 2700 °C
Density: 3.16
Storage Temperature: Ambient
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR