Silicon carbide, coarse powder 46 grit
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14470-0C
A14470-0E
A14470-36
AAAA14470-0CEA
456
USD
AAAA14470-0C
AAAA14470-0E
AAAA14470-36
Silicon carbide, coarse powder 46 grit
Silicon carbide
CAS No.: 409-21-2
Molecular Formula: SiC
Formula Weight: 40.10
Odor: Odorless
MDL No.: MFCD00049531
Formula:
SiC Melting Pt: 2700 °C Density: 3.16 Storage Temperature: Ambient |
MDL Number:
MFCD00049531 CAS Number: 409-21-2 EINECS: 206-991-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...