Iodoethane 98+% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with copper

A14444-0E A14444-22 A14444-36
AAAA14444-0EEA 584 USD
AAAA14444-0E AAAA14444-22 AAAA14444-36
Iodoethane 98+% stabilized
Iodoethane

Purity: 98+% CAS No.: 75-03-6 Molecular Formula: CH3CH2I Formula Weight: 155.97 Flash Point: 73°(163°F) Refractive Index: 1.5120 Melting Point: -108° Boiling Point: 70-73° Beilstein Reference: 505934 Merck Reference: 14,3813 MDL No.: MFCD00001091

Formula: C₂H₅I
Boiling Pt: 70…73 °C
Melting Pt: –108 °C
Density: 1.95
Flash Pt: >73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001091
CAS Number: 75-03-6
EINECS: 200-833-1
ADR: 6.1,III
Merck Index: 14,3813

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR