p-Toluenesulfonic acid monohydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate, 4-Methylbenzenesulfonic acid hydrate, Toluene-4-sulfonic acid monohydrate
A14119-0I
AAAA14119-36EA
75.48
USD
AAAA14119-36
AAAA14119-22
AAAA14119-0I
p-Toluenesulfonic acid monohydrate 98%
p-Toluenesulphonic acid monohydrate
Purity: 98%
CAS No.: 6192-52-5
Molecular Formula: C7H8O3S.H2O
Formula Weight: 190.22 (172.20anhy)
Flash Point: 150°(302°F)
Odor: Mild
Boiling Point: 140°/20mm
Beilstein Reference: 3568023
Merck Reference: 14,9533
UN No.: 2585
MDL No.: MFCD00142137
Formula:
C₇H₈O₃S·1H₂O Boiling Pt: 140 °C/20 mm Melting Pt: 103…106 °C (anhydr.) Density: 1.24 Flash Pt: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00142137 CAS Number: 6192-52-5 EINECS: 203-180-0 UN: 2585 ADR: 8,III Merck Index: 14,9533 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...