Tetrabutylammonium hydrogen sulfate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulfate, Tetrabutylammonium bisulfate, Tetrabutylammonium hydrogensulphate

A14047-0B A14047-18 A14047-30
AAAA14047-0BEA 371 USD
AAAA14047-0B AAAA14047-18 AAAA14047-30
Tetrabutylammonium hydrogen sulfate 97%
Tetrabutylammonium hydrogen sulphate

Phase transfer catalyst


Purity: 97% CAS No.: 32503-27-8 Molecular Formula: [CH3(CH2)3]4NHSO4 Formula Weight: 339.54 Melting Point: 168-172° MDL No.: MFCD00011637

Formula: (C₄H₉)₄NHSO₄
Melting Pt: 168…172 °C
Density: 1.01 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR