D-(-)-Mannitol ≥97.0%, white powder
Supplier: Thermo Scientific Chemicals
Synonyms:
Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
A14030-0B
A14030-30
A14030-0I
AAAA14030-0BEA
116.73
USD
AAAA14030-0B
AAAA14030-30
AAAA14030-0I
D-(-)-Mannitol ≥97.0%, white powder
D(-)-Mannitol
Purity: 99%
CAS No.: 69-65-8
Molecular Formula: C6H14O6
Formula Weight: 182.17
Odor: Odorless
Melting Point: 165-167°
Boiling Point: 290-295°/3.5mm
Beilstein Reference: 1721898
Merck Reference: 14,5745
Optical Rotation: +24° (in borax solution)
MDL No.: MFCD00064287
Formula:
HOH₂C(CH(OH))₄CH₂OH MW: 182.17 g/mol Boiling Pt: 290…295 °C (3.5 mmHg) Melting Pt: 165…167 °C Density: 1.489 Storage Temperature: Ambient |
MDL Number:
MFCD00064287 CAS Number: 69-65-8 EINECS: 200-711-8 Merck Index: 14,5745 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...