Copper(II) nitrate hemipentahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate

A14004-0B A14004-0I A14004-30
AAAA14004-0BEA 168.69 USD
AAAA14004-0B AAAA14004-0I AAAA14004-30
Copper(II) nitrate hemipentahydrate 98%
Copper (II) nitrate hemipentahydrate

Purity: 98% CAS No.: 19004-19-4 Molecular Formula: CuN2O6.2.5H2O Formula Weight: 232.59 (187.55anhy) Odor: Odorless Melting Point: ca 115° Boiling Point: 170° dec. Merck Reference: 14,2643 UN No.: 3085 MDL No.: MFCD00149671

Formula: Cu(NO₃)₂·2.5H₂O
Boiling Pt: 170 °C (dec.)
Melting Pt: ∼ 115 °C
Density: 2.32
Storage Temperature: Ambient
MDL Number: MFCD02253001
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,2643

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR