Potassium sulfate 99%

Supplier: Thermo Scientific Chemicals

A13975-0B A13975-0I
AAAA13975-0BEA 70.23 USD
AAAA13975-0B AAAA13975-0I
Potassium sulfate 99%
Potassium sulphate

Purity: 99% CAS No.: 7778-80-5 Molecular Formula: K2SO4 Formula Weight: 174.27 Odor: Odorless Melting Point: 1069° Boiling Point: 1689° Merck Reference: 14,7674 MDL No.: MFCD00011388

Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 2.662
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 14,7674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR